Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-methoxybenzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-methoxybenzoyl chloride
CAS 56658-04-9
Molecular Formula C8H6BrClO2
Formula Weight 249.5
MDL Number MFCD00219937
UN Number UN3265
Synonym 6-Bromo-m-anisoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1916403

  SDS
alfa aesar™
A1916403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1916409

  SDS
alfa aesar™
A1916409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.5
Assay 97%
Chemical Name or Material 2-Bromo-5-methoxybenzoyl chloride
MDL Number MFCD00219937
Quantity 1g
CAS 56658-04-9
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
UN Number UN3265
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C8H6BrClO2
Boiling Point 175° to 177°C (15mmHg)
Synonym 6-Bromo-m-anisoyl chloride
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.