Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-methoxybenzamide, 98%

$90.96 - $303.24

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-methoxybenzamide
CAS 99848-43-8
Molecular Formula C8H8BrNO2
Formula Weight 230.66
MDL Number MFCD00208336
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3125006

  SDS
alfa aesar™
H3125006
5g Each for $90.96

AAH3125014

  SDS
alfa aesar™
H3125014
25g Each for $303.24
Description & Specifications

Specifications

Formula Weight 230.66
CAS 99848-43-8
Quantity 5g
Chemical Name or Material 2-Bromo-5-methoxybenzamide
Assay 98%
Molecular Formula C8H8BrNO2
MDL Number MFCD00208336
Melting Point 159° to 161°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.