missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-iodotoluene, TCI America™

$58.06 - $172.64

Chemical Identifiers

CAS 202865-85-8
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.933
MDL Number MFCD00070749
InChI Key QSQOGKONVJDRNH-UHFFFAOYSA-N
Synonym 2-bromo-5-iodotoluene, benzene,1-bromo-4-iodo-2-methyl, benzene, 1-bromo-4-iodo-2-methyl, pubchem3791, 2-bromo-5-iodo toluene, 2-bromo-5-iodo-toluene, 2-bromo-5-iodotoluene;, acmc-1cq3e, ksc543o8j, 1-bromo-2-methyl-4-iodobenzene
PubChem CID 2735574
IUPAC Name 1-bromo-4-iodo-2-methylbenzene
SMILES CC1=C(C=CC(=C1)I)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30995G View Documents TCI America
B30995G
5g Each for $58.06
 
B309925G View Documents TCI America
B309925G
25g Each for $172.64
 
Specifications

Chemical Identifiers

202865-85-8
296.933
QSQOGKONVJDRNH-UHFFFAOYSA-N
2735574
CC1=C(C=CC(=C1)I)Br
C7H6BrI
MFCD00070749
2-bromo-5-iodotoluene, benzene,1-bromo-4-iodo-2-methyl, benzene, 1-bromo-4-iodo-2-methyl, pubchem3791, 2-bromo-5-iodo toluene, 2-bromo-5-iodo-toluene, 2-bromo-5-iodotoluene;, acmc-1cq3e, ksc543o8j, 1-bromo-2-methyl-4-iodobenzene
1-bromo-4-iodo-2-methylbenzene

Specifications

Yellow
202865-85-8
MFCD00070749
QSQOGKONVJDRNH-UHFFFAOYSA-N
1-bromo-4-iodo-2-methylbenzene
2735574
Clear Liquid at 20°C
2-Bromo-5-iodotoluene
5g
C7H6BrI
2-bromo-5-iodotoluene, benzene,1-bromo-4-iodo-2-methyl, benzene, 1-bromo-4-iodo-2-methyl, pubchem3791, 2-bromo-5-iodo toluene, 2-bromo-5-iodo-toluene, 2-bromo-5-iodotoluene;, acmc-1cq3e, ksc543o8j, 1-bromo-2-methyl-4-iodobenzene
CC1=C(C=CC(=C1)I)Br
296.933
296.93
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No