Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-iodotoluene, 98+%

$232.04 - $232.04

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-iodotoluene
CAS 202865-85-8
Molecular Formula C7H6BrI
Formula Weight 296.93
MDL Number MFCD00070749
Beilstein 8760525
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2246209

  SDS
alfa aesar™
B2246209
10g Each for $232.04
Description & Specifications

Specifications

Formula Weight 296.93
Quantity 10g
Molecular Formula C7H6BrI
Health Hazard 1 Warning
MDL Number MFCD00070749
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Bromo-5-iodotoluene
CAS 202865-85-8
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Beilstein 8760525
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.