missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-iodopyridine, TCI America™

$68.75 - $206.25

Chemical Identifiers

CAS 73290-22-9
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.894
MDL Number MFCD03095201
InChI Key LLKRSJVPTKFSLS-UHFFFAOYSA-N
Synonym 2-bromo-5-iodo-pyridine, pyridine, 2-bromo-5-iodo, 5-iodo-2-bromopyridine, pubchem5969, 5-iodo-2-bromo-pyridine, acmc-1bb8d, ksc493q0p, 2-bromo-5-iodopyridine
PubChem CID 4738271
IUPAC Name 2-bromo-5-iodopyridine
SMILES C1=CC(=NC=C1I)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B37655G View Documents TCI America
B37655G
5g Each for $68.75
 
B376525G View Documents TCI America
B376525G
25g Each for $206.25
 
Specifications

Chemical Identifiers

73290-22-9
283.894
LLKRSJVPTKFSLS-UHFFFAOYSA-N
4738271
C1=CC(=NC=C1I)Br
C5H3BrIN
MFCD03095201
2-bromo-5-iodo-pyridine, pyridine, 2-bromo-5-iodo, 5-iodo-2-bromopyridine, pubchem5969, 5-iodo-2-bromo-pyridine, acmc-1bb8d, ksc493q0p, 2-bromo-5-iodopyridine
2-bromo-5-iodopyridine

Specifications

White-Yellow
5g
C5H3BrIN
2-bromo-5-iodo-pyridine, pyridine, 2-bromo-5-iodo, 5-iodo-2-bromopyridine, pubchem5969, 5-iodo-2-bromo-pyridine, acmc-1bb8d, ksc493q0p, 2-bromo-5-iodopyridine
C1=CC(=NC=C1I)Br
283.894
283.89
≥97.0% (GC)
123°C
73290-22-9
MFCD03095201
LLKRSJVPTKFSLS-UHFFFAOYSA-N
2-bromo-5-iodopyridine
4738271
Crystal-Powder at 20°C
2-Bromo-5-iodopyridine
Safety and Handling

Safety and Handling

TSCA : No