missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-5-iodopyridine, 97%, ACROS Organics™
Chemical Identifiers
CAS | 73290-22-9 |
---|---|
Molecular Formula | C5H3BrIN |
Molecular Weight (g/mol) | 283.89 |
InChI Key | LLKRSJVPTKFSLS-UHFFFAOYSA-N |
Synonym | 2-bromo-5-iodo-pyridine, 2-bromo-5-iodopyridine, 5-iodo-2-bromo-pyridine, 5-iodo-2-bromopyridine, acmc-1bb8d, ksc493q0p, pubchem5969, pyridine, 2-bromo-5-iodo |
PubChem CID | 4738271 |
IUPAC Name | 2-bromo-5-iodopyridine |
SMILES | C1=CC(=NC=C1I)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC436850050
|
Thermo Fisher Scientific
AC436850050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC436850250
|
Thermo Fisher Scientific
AC436850250 |
25g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
73290-22-9 | |
283.89 | |
2-bromo-5-iodo-pyridine, 2-bromo-5-iodopyridine, 5-iodo-2-bromo-pyridine, 5-iodo-2-bromopyridine, acmc-1bb8d, ksc493q0p, pubchem5969, pyridine, 2-bromo-5-iodo | |
2-bromo-5-iodopyridine |
C5H3BrIN | |
LLKRSJVPTKFSLS-UHFFFAOYSA-N | |
4738271 | |
C1=CC(=NC=C1I)Br |
Specifications
Authentic | |
White | |
5g | |
96.0 | |
96% min. (GC) | |
2-bromo-5-iodo-pyridine, 2-bromo-5-iodopyridine, 5-iodo-2-bromo-pyridine, 5-iodo-2-bromopyridine, acmc-1bb8d, ksc493q0p, pubchem5969, pyridine, 2-bromo-5-iodo | |
C1=CC(=NC=C1I)Br | |
283.89 | |
283.89 | |
97% |
Glass bottle | |
123°C to 123.6°C | |
73290-22-9 | |
100.0 | |
C5H3BrIN | |
LLKRSJVPTKFSLS-UHFFFAOYSA-N | |
2-bromo-5-iodopyridine | |
4738271 | |
Powder | |
2-Bromo-5-iodopyridine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning