Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-iodopyridine, 97%

$33.76 - $130.64

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-iodopyridine
CAS 73290-22-9
Molecular Formula C5H3BrIN
Formula Weight 283.89
MDL Number MFCD03095201
Beilstein 109100
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2001603

  SDS
alfa aesar™
L2001603
1g Each for $33.76

AAL2001606

  SDS
alfa aesar™
L2001606
5g Each for $130.64
Description & Specifications

Specifications

Formula Weight 283.89
Chemical Name or Material 2-Bromo-5-iodopyridine
CAS 73290-22-9
Molecular Formula C5H3BrIN
MDL Number MFCD03095201
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Melting Point 123° to 124°C
Beilstein 109100
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.