missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-5-hydroxypyrazine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC435890050
Specifications
2-Bromo-5-hydroxypyrazine | |
Glass bottle | |
115°C to 119°C | |
374063-92-0 | |
100.0 | |
C4H3BrN2O | |
2 1h-pyrazinone, 5-bromo, 2-bromo-5-hydroxy-pyrazine, 2-bromo-5-hydroxypyrazine, 2-hydroxy-5-bromopyrazine, 5-bromo-2-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 5-bromopyrazin-2-ol, acmc-20a0of, pubchem7870 | |
C1=C(N=CC(=O)N1)Br | |
174.98 | |
174.98 | |
97% |
Authentic | |
White to Brown | |
5g | |
96.0 | |
96% min. (HPLC) | |
MFCD06245330 | |
ITTXBHQAWOFJAI-UHFFFAOYSA-N | |
5-bromo-1H-pyrazin-2-one | |
2771618 | |
Crystalline Powder |
Chemical Identifiers
374063-92-0 | |
174.98 | |
ITTXBHQAWOFJAI-UHFFFAOYSA-N | |
2771618 | |
C1=C(N=CC(=O)N1)Br |
C4H3BrN2O | |
MFCD06245330 | |
2 1h-pyrazinone, 5-bromo, 2-bromo-5-hydroxy-pyrazine, 2-bromo-5-hydroxypyrazine, 2-hydroxy-5-bromopyrazine, 5-bromo-2-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 5-bromopyrazin-2-ol, acmc-20a0of, pubchem7870 | |
5-bromo-1H-pyrazin-2-one |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap
GHS Signal Word: Warning