Learn More
2-Bromo-5-hydroxybenzaldehyde, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6488422
Description
2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.
Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
2-Bromo-5-hydroxybenzaldehyde | |
130°C to 135°C | |
MFCD03033065 | |
UN3077 | |
4-bromo-3-formylphenol, 2-bromo-5-hydroxy-benzaldehyde, 2-bromo-5-hydroxy benzaldehyde, benzaldehyde, 2-bromo-5-hydroxy, 2-bromo-5-hydroxybenzadehyde, benzaldehyde,2-bromo-5-hydroxy, pubchem14430, acmc-209haw, 6-brom-3-oxy-benzaldehyd | |
SCRQAWQJSSKCFN-UHFFFAOYSA-N | |
2-bromo-5-hydroxybenzaldehyde | |
387179 | |
95% |
2973-80-0 | |
C7H5BrO2 | |
100 g | |
2079921 | |
Soluble in chloroform,dichloromethane and ethyl acetate. | |
OC1=CC(C=O)=C(Br)C=C1 | |
201.02 | |
201.02 | |
Solid |
Chemical Identifiers
2973-80-0 | |
201.02 | |
SCRQAWQJSSKCFN-UHFFFAOYSA-N | |
2-bromo-5-hydroxybenzaldehyde |
C7H5BrO2 | |
MFCD03033065 | |
387179 | |
OC1=CC(C=O)=C(Br)C=C1 |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only