missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-hydroxy-3-methylpyridine, 97%

$147.27 - $467.26

Chemical Identifiers

CAS 1003711-43-0
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD09839267
InChI Key KCCYUVIIKCPVDH-UHFFFAOYSA-N
Synonym 2-bromo-5-hydroxy-3-methylpyridine, 2-bromo-5-hydroxy-3-picoline, pubchem5966, 6-bromo-5-methyl-3-pyridinol, 3-pyridinol,6-bromo-5-methyl, 6-bromo-5-methyl-pyridin-3-ol, 6-bromanyl-5-methyl-pyridin-3-ol, abbypharma ap-30-3129
PubChem CID 29919363
IUPAC Name 6-bromo-5-methylpyridin-3-ol
SMILES CC1=CC(=CN=C1Br)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64567MD View Documents Alfa Aesar™
H64567MD
250mg Each for $147.27
 
AAH6456703 View Documents Alfa Aesar™
H6456703
1g Each for $467.26
 
Specifications

Chemical Identifiers

1003711-43-0
188.024
KCCYUVIIKCPVDH-UHFFFAOYSA-N
29919363
CC1=CC(=CN=C1Br)O
C6H6BrNO
MFCD09839267
2-bromo-5-hydroxy-3-methylpyridine, 2-bromo-5-hydroxy-3-picoline, pubchem5966, 6-bromo-5-methyl-3-pyridinol, 3-pyridinol,6-bromo-5-methyl, 6-bromo-5-methyl-pyridin-3-ol, 6-bromanyl-5-methyl-pyridin-3-ol, abbypharma ap-30-3129
6-bromo-5-methylpyridin-3-ol

Specifications

White
1003711-43-0
MFCD09839267
KCCYUVIIKCPVDH-UHFFFAOYSA-N
6-bromo-5-methylpyridin-3-ol
29919363
Powder
2-Bromo-5-hydroxy-3-methylpyridine
250mg
C6H6BrNO
2-bromo-5-hydroxy-3-methylpyridine, 2-bromo-5-hydroxy-3-picoline, pubchem5966, 6-bromo-5-methyl-3-pyridinol, 3-pyridinol,6-bromo-5-methyl, 6-bromo-5-methyl-pyridin-3-ol, 6-bromanyl-5-methyl-pyridin-3-ol, abbypharma ap-30-3129
CC1=CC(=CN=C1Br)O
188.024
188.02
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning