Please login to your online account to display your discounted pricing

2-Bromo-5-fluorotoluene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-fluorotoluene
Name Note 99%
CAS 452-63-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H6BrF
Linear Formula BrC6H3(F)CH3
Formula Weight 189.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC294040050

  SDS
thermo fisher scientific
294040050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 189.03
Quantity 5g
Color Clear colorless
TSCA TSCA
EINECS Number 207-203-5
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00017921
Refractive Index 1.5270 to 1.5290 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Chemical Name or Material 2-Bromo-5-fluorotoluene
Molecular Formula C7H6BrF
CAS Min % 98.5
Name Note 99%
CAS 452-63-1
Assay Percent Range 99%
Linear Formula BrC6H3(F)CH3
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.