Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-fluoropyridine, 98%

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-fluoropyridine
CAS 41404-58-4
Molecular Formula C5H3BrFN
Formula Weight 175.99
MDL Number MFCD00234011
UN Number UN2811
Beilstein 1561456
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1953603

  SDS
alfa aesar™
L1953603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1953606

  SDS
alfa aesar™
L1953606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.99
CAS 41404-58-4
Chemical Name or Material 2-Bromo-5-fluoropyridine
Assay 98%
Quantity 1g
Flash Point 73°C (163°F)
Molecular Formula C5H3BrFN
MDL Number MFCD00234011
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 26° to 31°C
Boiling Point 80° to 83°C (44mmHg)
Beilstein 1561456
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.