Learn More
2-Bromo-5-fluorophenol, 97%, ACROS Organics™
$68.54 - $68.54
Chemical Identifiers
CAS | 147460-41-1 |
---|---|
Molecular Formula | C6H4BrFO |
Molecular Weight (g/mol) | 191 |
MDL Number | MFCD00040939 |
InChI Key | HUVAOAVBKOVPBZ-UHFFFAOYSA-N |
Synonym | 2-bromo-5-fluoro-phenol, 2-bromo-5-fluorophenol, 2-bromo-5-fluorphenol, 2-bromo-5-fluorphenol;, acmc-209cyi, attercop-chm at106843, ksc493i7h, paragos 390207, phenol, 2-bromo-5-fluoro, pubchem1964 |
PubChem CID | 2724600 |
IUPAC Name | 2-bromo-5-fluorophenol |
SMILES | C1=CC(=C(C=C1F)O)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC329490010
|
Thermo Fisher Scientific
AC329490010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC329490050
|
Thermo Fisher Scientific
329490050 |
5g | Glass bottle |
Each for $68.54
|
|
AC329490250
|
Thermo Fisher Scientific
AC329490250 |
25g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
147460-41-1 | |
191 | |
HUVAOAVBKOVPBZ-UHFFFAOYSA-N | |
2724600 | |
C1=CC(=C(C=C1F)O)Br |
C6H4BrFO | |
MFCD00040939 | |
2-bromo-5-fluoro-phenol, 2-bromo-5-fluorophenol, 2-bromo-5-fluorphenol, 2-bromo-5-fluorphenol;, acmc-209cyi, attercop-chm at106843, ksc493i7h, paragos 390207, phenol, 2-bromo-5-fluoro, pubchem1964 | |
2-bromo-5-fluorophenol |
Specifications
1.7170g/mL | |
Authentic | |
1.5520 to 1.5540 | |
89.0°C (1.0 mmHg) | |
42.0°C to 44.0°C | |
147460-41-1 | |
100.0 | |
C6H4BrFO | |
MFCD00040939 | |
HUVAOAVBKOVPBZ-UHFFFAOYSA-N | |
2-bromo-5-fluorophenol | |
2724600 | |
Liquid | |
2-Bromo-5-fluorophenol, 97% |
87°C | |
Glass bottle | |
1.717 | |
Colorless to Yellow | |
1g | |
96.0 | |
96% min. (GC) | |
BrC6H3(F)OH | |
2-bromo-5-fluoro-phenol, 2-bromo-5-fluorophenol, 2-bromo-5-fluorphenol, 2-bromo-5-fluorphenol;, acmc-209cyi, attercop-chm at106843, ksc493i7h, paragos 390207, phenol, 2-bromo-5-fluoro, pubchem1964 | |
C1=CC(=C(C=C1F)O)Br | |
191 | |
191 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA