missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-fluorophenol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC329490010

 View more versions of this product

Catalog No. AC329490010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Bromo-5-fluorophenol
87°C
Glass bottle
1.717
Colorless to Yellow
1g
147460-41-1
100.0
C6H4BrFO
MFCD00040939
HUVAOAVBKOVPBZ-UHFFFAOYSA-N
2-bromo-5-fluorophenol
2724600
Liquid
1.7170g/mL
Authentic
1.5520 to 1.5540
89.0°C (1.0 mmHg)
42.0°C to 44.0°C
97%
96.0
96% min. (GC)
BrC6H3(F)OH
2-bromo-5-fluoro-phenol, 2-bromo-5-fluorophenol, 2-bromo-5-fluorphenol, 2-bromo-5-fluorphenol;, acmc-209cyi, attercop-chm at106843, ksc493i7h, paragos 390207, phenol, 2-bromo-5-fluoro, pubchem1964
C1=CC(=C(C=C1F)O)Br
191
191
97%

Chemical Identifiers

147460-41-1
191
HUVAOAVBKOVPBZ-UHFFFAOYSA-N
2724600
C1=CC(=C(C=C1F)O)Br
C6H4BrFO
MFCD00040939
2-bromo-5-fluoro-phenol, 2-bromo-5-fluorophenol, 2-bromo-5-fluorphenol, 2-bromo-5-fluorphenol;, acmc-209cyi, attercop-chm at106843, ksc493i7h, paragos 390207, phenol, 2-bromo-5-fluoro, pubchem1964
2-bromo-5-fluorophenol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents