missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-fluorophenol, 97%

Manufacturer:  Alfa Aesar™ A1506422

 View more versions of this product

Catalog No. AAA1506422



Specifications

Specifications

2-Bromo-5-fluorophenol
184°C to 186°C
147460-41-1
MFCD00040939
phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843
C1=CC(=C(C=C1F)O)Br
190.999
191
87°C (188°F)
100g
C6H4BrFO
7915214
HUVAOAVBKOVPBZ-UHFFFAOYSA-N
2-bromo-5-fluorophenol
2724600
97%

Chemical Identifiers

147460-41-1
190.999
HUVAOAVBKOVPBZ-UHFFFAOYSA-N
2724600
C1=CC(=C(C=C1F)O)Br
C6H4BrFO
MFCD00040939
phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843
2-bromo-5-fluorophenol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents