Learn More
2-Bromo-5-fluorophenol, 97%, Thermo Scientific Chemicals
$78.55 - $247.14
Chemical Identifiers
CAS | 147460-41-1 |
---|---|
Molecular Formula | C6H4BrFO |
Molecular Weight (g/mol) | 190.999 |
MDL Number | MFCD00040939 |
InChI Key | HUVAOAVBKOVPBZ-UHFFFAOYSA-N |
Synonym | phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843 |
PubChem CID | 2724600 |
IUPAC Name | 2-bromo-5-fluorophenol |
SMILES | C1=CC(=C(C=C1F)O)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1506406
|
Thermo Scientific Chemicals
A1506406 |
5 g |
Each for $78.55
|
|
|||||
AAA1506414
|
Thermo Scientific Chemicals
A1506414 |
25 g |
Each for $247.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
147460-41-1 | |
190.999 | |
HUVAOAVBKOVPBZ-UHFFFAOYSA-N | |
2724600 | |
C1=CC(=C(C=C1F)O)Br |
C6H4BrFO | |
MFCD00040939 | |
phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843 | |
2-bromo-5-fluorophenol |
Specifications
147460-41-1 | |
184°C to 186°C | |
C6H4BrFO | |
MFCD00040939 | |
7915214 | |
HUVAOAVBKOVPBZ-UHFFFAOYSA-N | |
2-bromo-5-fluorophenol | |
2724600 | |
97% |
1.717 | |
87°C (188°F) | |
1.554 | |
5 g | |
phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843 | |
C1=CC(=C(C=C1F)O)Br | |
190.999 | |
191 | |
2-Bromo-5-fluorophenol |
Safety and Handling
GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only