missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-fluorobenzotrifluoride, TCI America™

$64.17 - $192.50

Chemical Identifiers

CAS 40161-55-5
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD00040937
InChI Key AIDVAZGOACECLJ-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorobenzotrifluoride, 1-bromo-4-fluoro-2-trifluoromethyl benzene, 1-bromo-2-trifluoromethyl-4-fluorobenzene, 4-fluoro-2-trifluoromethyl bromobenzene, 2-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, benzene, 1-bromo-4-fluoro-2-trifluoromethyl, 2-bromo-5-fluoro benzotrifluoride, 2-bromo-5-fluorobenzotrifluoride, pubchem1637, pubchem1643
PubChem CID 520983
IUPAC Name 1-bromo-4-fluoro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1F)C(F)(F)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28005G View Documents TCI America
B28005G
5g Each for $64.17
 
B280025G View Documents TCI America
B280025G
25g Each for $192.50
 
Specifications

Chemical Identifiers

40161-55-5
242.999
AIDVAZGOACECLJ-UHFFFAOYSA-N
520983
C1=CC(=C(C=C1F)C(F)(F)F)Br
C7H3BrF4
MFCD00040937
2-bromo-5-fluorobenzotrifluoride, 1-bromo-4-fluoro-2-trifluoromethyl benzene, 1-bromo-2-trifluoromethyl-4-fluorobenzene, 4-fluoro-2-trifluoromethyl bromobenzene, 2-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, benzene, 1-bromo-4-fluoro-2-trifluoromethyl, 2-bromo-5-fluoro benzotrifluoride, 2-bromo-5-fluorobenzotrifluoride, pubchem1637, pubchem1643
1-bromo-4-fluoro-2-(trifluoromethyl)benzene

Specifications

143°C
5g
C7H3BrF4
2-bromo-5-fluorobenzotrifluoride, 1-bromo-4-fluoro-2-trifluoromethyl benzene, 1-bromo-2-trifluoromethyl-4-fluorobenzene, 4-fluoro-2-trifluoromethyl bromobenzene, 2-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, benzene, 1-bromo-4-fluoro-2-trifluoromethyl, 2-bromo-5-fluoro benzotrifluoride, 2-bromo-5-fluorobenzotrifluoride, pubchem1637, pubchem1643
C1=CC(=C(C=C1F)C(F)(F)F)Br
242.999
243.00
≥97.0% (GC)
Yellow
40161-55-5
MFCD00040937
AIDVAZGOACECLJ-UHFFFAOYSA-N
1-bromo-4-fluoro-2-(trifluoromethyl)benzene
520983
Clear Liquid at 20°C
2-Bromo-5-fluorobenzotrifluoride
Safety and Handling

Safety and Handling

TSCA : No