missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-fluorobenzotrifluoride, 97%, ACROS Organics™

Chemical Identifiers

CAS 40161-55-5
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243
InChI Key AIDVAZGOACECLJ-UHFFFAOYSA-N
Synonym 1-bromo-4-fluoro-3-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 2-fluoro-5-bromobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-2-fluoro-trifluoromethyl benzene, 5-bromo-2-fluorobenzotrifluoride, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, acmc-209j4s, benzene, 4-bromo-1-fluoro-2-trifluoromethyl
PubChem CID 2736325
IUPAC Name 4-bromo-1-fluoro-2-(trifluoromethyl)benzene
SMILES FC1=CC(=C(Br)C=C1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC442570010
SDS View Documents
Thermo Fisher Scientific
AC442570010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC442570050
SDS View Documents
Thermo Fisher Scientific
AC442570050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

40161-55-5
243
1-bromo-4-fluoro-3-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 2-fluoro-5-bromobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-2-fluoro-trifluoromethyl benzene, 5-bromo-2-fluorobenzotrifluoride, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, acmc-209j4s, benzene, 4-bromo-1-fluoro-2-trifluoromethyl
4-bromo-1-fluoro-2-(trifluoromethyl)benzene
C7H3BrF4
AIDVAZGOACECLJ-UHFFFAOYSA-N
2736325
FC1=CC(=C(Br)C=C1)C(F)(F)F

Specifications

1.7500g/mL
Authentic
1.75
Colorless to Yellow
40161-55-5
100.0
C7H3BrF4
AIDVAZGOACECLJ-UHFFFAOYSA-N
4-bromo-1-fluoro-2-(trifluoromethyl)benzene
2736325
Liquid
2-Bromo-5-fluorobenzotrifluoride
63°C
Glass bottle
136°C to 143°C
1g
96.0
96% min. (GC)
1-bromo-4-fluoro-3-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 2-fluoro-5-bromobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-2-fluoro-trifluoromethyl benzene, 5-bromo-2-fluorobenzotrifluoride, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, acmc-209j4s, benzene, 4-bromo-1-fluoro-2-trifluoromethyl
FC1=CC(=C(Br)C=C1)C(F)(F)F
243
243
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at

GHS Signal Word: Warning

SDS