missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-5-fluorobenzotrifluoride, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442570050
Specifications
2-Bromo-5-fluorobenzotrifluoride | |
63°C | |
Glass bottle | |
136°C to 143°C | |
5g | |
96.0 | |
96% min. (GC) | |
1-bromo-4-fluoro-3-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 2-fluoro-5-bromobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-2-fluoro-trifluoromethyl benzene, 5-bromo-2-fluorobenzotrifluoride, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, acmc-209j4s, benzene, 4-bromo-1-fluoro-2-trifluoromethyl | |
FC1=CC(=C(Br)C=C1)C(F)(F)F | |
243 | |
243 | |
97% |
1.7500g/mL | |
Authentic | |
1.75 | |
Colorless to Yellow | |
40161-55-5 | |
100.0 | |
C7H3BrF4 | |
AIDVAZGOACECLJ-UHFFFAOYSA-N | |
4-bromo-1-fluoro-2-(trifluoromethyl)benzene | |
2736325 | |
Liquid |
Chemical Identifiers
40161-55-5 | |
243 | |
1-bromo-4-fluoro-3-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 2-fluoro-5-bromobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-2-fluoro-trifluoromethyl benzene, 5-bromo-2-fluorobenzotrifluoride, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, acmc-209j4s, benzene, 4-bromo-1-fluoro-2-trifluoromethyl | |
4-bromo-1-fluoro-2-(trifluoromethyl)benzene |
C7H3BrF4 | |
AIDVAZGOACECLJ-UHFFFAOYSA-N | |
2736325 | |
FC1=CC(=C(Br)C=C1)C(F)(F)F |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at
GHS Signal Word: Warning