Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-fluorobenzotrifluoride, 97%

$482.16 - $482.16

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-fluorobenzotrifluoride
CAS 40161-55-5
Molecular Formula C7H3BrF4
Formula Weight 243
MDL Number MFCD00040937
Beilstein 2643544
Synonym 1-Bromo-4-fluoro-2-(trifluoromethyl)benzene; 4-Fluoro-2-(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2400514

  SDS
alfa aesar™
B2400514
25g Each for $482.16

AAB2400503

  SDS
alfa aesar™
B2400503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2400506

  SDS
alfa aesar™
B2400506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Bromo-5-fluorobenzotrifluoride
CAS 40161-55-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C7H3BrF4
Formula Weight 243
MDL Number MFCD00040937
Beilstein 2643544
Synonym 1-Bromo-4-fluoro-2-(trifluoromethyl)benzene; 4-Fluoro-2-(trifluoromethyl)bromobenzene
Boiling Point 136°C
Quantity 25g
Flash Point 63°C (145°F)
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.