missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-5-fluorobenzoic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2539414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromo-5-fluorobenzoic acid | |
154°C to 158°C | |
MFCD00142874 | |
2575978 | |
OQBMJMJZMDBQSM-UHFFFAOYSA-M | |
2-bromo-5-fluorobenzoic acid | |
2778181 | |
≥98% |
394-28-5 | |
C7H3BrFO2 | |
25 g | |
benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747 | |
[O-]C(=O)C1=CC(F)=CC=C1Br | |
218.00 | |
219.01 |
Chemical Identifiers
394-28-5 | |
218.00 | |
OQBMJMJZMDBQSM-UHFFFAOYSA-M | |
2778181 | |
[O-]C(=O)C1=CC(F)=CC=C1Br |
C7H3BrFO2 | |
MFCD00142874 | |
benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747 | |
2-bromo-5-fluorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : Yes
RUO – Research Use Only