missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-fluorobenzamide, 96%

$60.92 - $220.53

Chemical Identifiers

CAS 1006-34-4
Molecular Formula C7H5BrFNO
Molecular Weight (g/mol) 218.025
MDL Number MFCD03094322
InChI Key OCKACIBALIJPNT-UHFFFAOYSA-N
Synonym acmc-1br05, benzamide,2-bromo-5-fluoro, benzamide, 2-bromo-5-fluoro
PubChem CID 2778464
IUPAC Name 2-bromo-5-fluorobenzamide
SMILES C1=CC(=C(C=C1F)C(=O)N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62366MD View Documents Alfa Aesar™
H62366MD
250mg Each for $60.92
 
AAH6236603 View Documents Alfa Aesar™
H6236603
1g Each for $220.53
 
Specifications

Chemical Identifiers

1006-34-4
218.025
OCKACIBALIJPNT-UHFFFAOYSA-N
2778464
C1=CC(=C(C=C1F)C(=O)N)Br
C7H5BrFNO
MFCD03094322
acmc-1br05, benzamide,2-bromo-5-fluoro, benzamide, 2-bromo-5-fluoro
2-bromo-5-fluorobenzamide

Specifications

250mg
C7H5BrFNO
acmc-1br05, benzamide,2-bromo-5-fluoro, benzamide, 2-bromo-5-fluoro
C1=CC(=C(C=C1F)C(=O)N)Br
218.025
218.02
2-Bromo-5-fluorobenzamide
1006-34-4
MFCD03094322
OCKACIBALIJPNT-UHFFFAOYSA-N
2-bromo-5-fluorobenzamide
2778464
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning