missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-fluorobenzaldehyde, 96%, ACROS Organics™

Chemical Identifiers

CAS 94569-84-3
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C7H4BrFO
Linear Formula FC6H3(Br)CHO
Formula Weight 203.01
MDL Number MFCD00142872
Chemical Name or Material 2-Bromo-5-fluorobenzaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC398910050 SDS Thermo Fisher Scientific
AC398910050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398910010 SDS Thermo Fisher Scientific
AC398910010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

94569-84-3
100.0
C7H4BrFO
203.01
2-Bromo-5-fluorobenzaldehyde
95.0
96%
FC6H3(Br)CHO
MFCD00142872

Specifications

94569-84-3
100.0
96%
C7H4BrFO
203.01
2-Bromo-5-fluorobenzaldehyde
95.0
Glass bottle
5g
FC6H3(Br)CHO
MFCD00142872

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.