Please login to your online account to display your discounted pricing

2-Bromo-5-fluoroaniline, 99%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC398840250

 View more versions of this product

Catalog No. AC398840250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 25g
Chemical Name or Material 2-Bromo-5-fluoroaniline
Assay Percent Range 99%
Molecular Formula C6H5BrFN
Linear Formula BrC6H3(F)NH2
MDL Number MFCD00070750
CAS 1003-99-2

Specifications

Chemical Name or Material 2-Bromo-5-fluoroaniline
CAS 1003-99-2
Assay Percent Range 99%
Molecular Formula C6H5BrFN
Linear Formula BrC6H3(F)NH2
MDL Number MFCD00070750