Please login to your online account to display your discounted pricing

2-Bromo-5-fluoroaniline, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-fluoroaniline
CAS 1003-99-2
Assay Percent Range 99%
Molecular Formula C6H5BrFN
Linear Formula BrC6H3(F)NH2
MDL Number MFCD00070750
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC398840050

 
thermo fisher scientific
AC398840050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC398840250

 
thermo fisher scientific
AC398840250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
CAS 1003-99-2
Chemical Name or Material 2-Bromo-5-fluoroaniline
Assay Percent Range 99%
Molecular Formula C6H5BrFN
Linear Formula BrC6H3(F)NH2
MDL Number MFCD00070750