missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-fluoro-3-nitropyridine, 95%

$141.42 - $422.54

Chemical Identifiers

CAS 652160-72-0
Molecular Formula C5H2BrFN2O2
Molecular Weight (g/mol) 220.985
MDL Number MFCD05662387
InChI Key XJFDIIHIXNIWQH-UHFFFAOYSA-N
Synonym 2-bromo-3-nitro-5-fluoropyridine, 2-bromo-5-fluoro-2-nitropyrdine, 2-bromo-5-fluoro-3-nitro-pyridine, pyridine,2-bromo-5-fluoro-3-nitro, pyridine, 2-bromo-5-fluoro-3-nitro, pubchem14284, 2-bromo-3-nitro-5-fluoro pyridine, 2-bromanyl-5-fluoranyl-3-nitro-pyridine
PubChem CID 20791243
IUPAC Name 2-bromo-5-fluoro-3-nitropyridine
SMILES C1=C(C(=NC=C1F)Br)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64208MD View Documents Alfa Aesar™
H64208MD
250mg Each for $141.42
 
AAH6420803 View Documents Alfa Aesar™
H6420803
1g Each for $422.54
 
Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

652160-72-0
220.985
XJFDIIHIXNIWQH-UHFFFAOYSA-N
20791243
C1=C(C(=NC=C1F)Br)[N+](=O)[O-]
C5H2BrFN2O2
MFCD05662387
2-bromo-3-nitro-5-fluoropyridine, 2-bromo-5-fluoro-2-nitropyrdine, 2-bromo-5-fluoro-3-nitro-pyridine, pyridine,2-bromo-5-fluoro-3-nitro, pyridine, 2-bromo-5-fluoro-3-nitro, pubchem14284, 2-bromo-3-nitro-5-fluoro pyridine, 2-bromanyl-5-fluoranyl-3-nitro-pyridine
2-bromo-5-fluoro-3-nitropyridine

Specifications

Orange
652160-72-0
MFCD05662387
2-bromo-3-nitro-5-fluoropyridine, 2-bromo-5-fluoro-2-nitropyrdine, 2-bromo-5-fluoro-3-nitro-pyridine, pyridine,2-bromo-5-fluoro-3-nitro, pyridine, 2-bromo-5-fluoro-3-nitro, pubchem14284, 2-bromo-3-nitro-5-fluoro pyridine, 2-bromanyl-5-fluoranyl-3-nitro-pyridine
C1=C(C(=NC=C1F)Br)[N+](=O)[O-]
220.985
220.99
95%
250mg
C5H2BrFN2O2
14253146
XJFDIIHIXNIWQH-UHFFFAOYSA-N
2-bromo-5-fluoro-3-nitropyridine
20791243
Solid
2-Bromo-5-fluoro-3-nitropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning