missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-decylthiophene, TCI America™

$409.45 - $1,228.36

Chemical Identifiers

CAS 514188-72-8
Molecular Formula C14H23BrS
Molecular Weight (g/mol) 303.302
InChI Key BLGUNQPKOOEOHU-UHFFFAOYSA-N
PubChem CID 58892816
IUPAC Name 2-bromo-5-decylthiophene
SMILES CCCCCCCCCCC1=CC=C(S1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B54061G View Documents TCI America
B54061G
1g Each for $409.45
 
B54065G View Documents TCI America
B54065G
5g Each for $1,228.36
 
Specifications

Chemical Identifiers

514188-72-8
303.302
58892816
CCCCCCCCCCC1=CC=C(S1)Br
C14H23BrS
BLGUNQPKOOEOHU-UHFFFAOYSA-N
2-bromo-5-decylthiophene

Specifications

135°C
1g
C14H23BrS
CCCCCCCCCCC1=CC=C(S1)Br
303.302
303.30
≥97.0% (GC)
Yellow
514188-72-8
BLGUNQPKOOEOHU-UHFFFAOYSA-N
2-bromo-5-decylthiophene
58892816
Clear Liquid at 20°C
2-Bromo-5-decylthiophene
Safety and Handling

Safety and Handling

TSCA : No