missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-cyanopyridine 97.0+%, TCI America™

$57.61 - $187.97

Chemical Identifiers

CAS 139585-70-9
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00234141
InChI Key XHYGUDGTUJPSNX-UHFFFAOYSA-N
Synonym 2-bromo-5-cyanopyridine, 6-bromonicotinonitrile, 2-bromo-5-pyridinecarbonitrile, 5-cyano-2-bromopyridine, 6-bromo-nicotinonitrile, 6-bromo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 6-bromo, pubchem5963, 2-bromo-5-cyano-pyridine, acmc-209cj9
PubChem CID 5005718
IUPAC Name 6-bromopyridine-3-carbonitrile
SMILES C1=CC(=NC=C1C#N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B40391G View Documents Tci America
B40391G
1g Each for $57.61
 
B40395G View Documents Tci America
B40395G
5g Each for $187.97
 
Specifications

Chemical Identifiers

139585-70-9
183.008
XHYGUDGTUJPSNX-UHFFFAOYSA-N
5005718
C1=CC(=NC=C1C#N)Br
C6H3BrN2
MFCD00234141
2-bromo-5-cyanopyridine, 6-bromonicotinonitrile, 2-bromo-5-pyridinecarbonitrile, 5-cyano-2-bromopyridine, 6-bromo-nicotinonitrile, 6-bromo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 6-bromo, pubchem5963, 2-bromo-5-cyano-pyridine, acmc-209cj9
6-bromopyridine-3-carbonitrile

Specifications

Undesignated
1g
C6H3BrN2
3439
XHYGUDGTUJPSNX-UHFFFAOYSA-N
6-bromopyridine-3-carbonitrile
5005718
Crystal-Powder at 20°C
2-Bromo-5-cyanopyridine
145°C
139585-70-9
MFCD00234141
2-bromo-5-cyanopyridine, 6-bromonicotinonitrile, 2-bromo-5-pyridinecarbonitrile, 5-cyano-2-bromopyridine, 6-bromo-nicotinonitrile, 6-bromo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 6-bromo, pubchem5963, 2-bromo-5-cyano-pyridine, acmc-209cj9
C1=CC(=NC=C1C#N)Br
183.008
183.01
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No