missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-cyanopyridine, 97%

$50.47 - $199.30

Chemical Identifiers

CAS 139585-70-9
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00234141
InChI Key XHYGUDGTUJPSNX-UHFFFAOYSA-N
Synonym 2-bromo-5-cyano-pyridine
PubChem CID 5005718
IUPAC Name 6-bromopyridine-3-carbonitrile
SMILES C1=CC(=NC=C1C#N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6617703 View Documents Alfa Aesar™
H6617703
1g Each for $50.47
 
AAH6617706 View Documents Alfa Aesar™
H6617706
5g Each for $199.30
 
Specifications

Chemical Identifiers

139585-70-9
183.008
XHYGUDGTUJPSNX-UHFFFAOYSA-N
5005718
C1=CC(=NC=C1C#N)Br
C6H3BrN2
MFCD00234141
2-bromo-5-cyano-pyridine
6-bromopyridine-3-carbonitrile

Specifications

1g
C6H3BrN2
UN3439
2-bromo-5-cyano-pyridine
C1=CC(=NC=C1C#N)Br
183.008
183.01
97%
139585-70-9
MFCD00234141
8760368
XHYGUDGTUJPSNX-UHFFFAOYSA-N
6-bromopyridine-3-carbonitrile
5005718
Solid
2-Bromo-5-cyanopyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures