Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-chlorotoluene, 98%

$36.01 - $262.18

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-chlorotoluene
CAS 14495-51-3
Molecular Formula C7H6BrCl
Formula Weight 205.48
MDL Number MFCD00000602
Beilstein 2500157
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1413218

 
alfa aesar™
A1413218
50g Each for $63.28

AAA1413214

 
alfa aesar™
A1413214
25g Each for $36.01

AAA1413230

 
alfa aesar™
A1413230
250g Each for $262.18
Description & Specifications

Specifications

Formula Weight 205.48
Chemical Name or Material 2-Bromo-5-chlorotoluene
Assay 98%
Molecular Formula C7H6BrCl
MDL Number MFCD00000602
CAS 14495-51-3
Beilstein 2500157
Boiling Point 98° to 99°C (25mmHg)
Quantity 50g
Flash Point 95°C (203°F)
Odor Aromatic
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.