missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-chloropyridine, 98%

$72.93 - $288.38

Chemical Identifiers

CAS 40473-01-6
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD00234006
InChI Key BZUUVQCSPHPUQA-UHFFFAOYSA-N
Synonym 5-chloro-2-bromopyridine, 2-bromo-5-chloro-pyridine, 2-brom-5-chlorpyridin, pyridine, 2-bromo-5-chloro, pubchem1169, acmc-209jeb, 2-bromo-5-chloro pyridine, ksc238i0d, tpc-py102, 2-bromanyl-5-chloranyl-pyridine
PubChem CID 817098
IUPAC Name 2-bromo-5-chloropyridine
SMILES C1=CC(=NC=C1Cl)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6436714 View Documents Alfa Aesar™
H6436714
25g Each for $72.93
 
AAH6436722 View Documents Alfa Aesar™
H6436722
100g Each for $288.38
 
Description

Description

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in the synthesis of novel halopyridinylboronic acids and esters.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

40473-01-6
192.44
BZUUVQCSPHPUQA-UHFFFAOYSA-N
817098
C1=CC(=NC=C1Cl)Br
C5H3BrClN
MFCD00234006
5-chloro-2-bromopyridine, 2-bromo-5-chloro-pyridine, 2-brom-5-chlorpyridin, pyridine, 2-bromo-5-chloro, pubchem1169, acmc-209jeb, 2-bromo-5-chloro pyridine, ksc238i0d, tpc-py102, 2-bromanyl-5-chloranyl-pyridine
2-bromo-5-chloropyridine

Specifications

128°C (16mmHg)
25g
C5H3BrClN
109098
BZUUVQCSPHPUQA-UHFFFAOYSA-N
2-bromo-5-chloropyridine
817098
Solid
2-Bromo-5-chloropyridine
65°C to 69°C (literature)
40473-01-6
MFCD00234006
5-chloro-2-bromopyridine, 2-bromo-5-chloro-pyridine, 2-brom-5-chlorpyridin, pyridine, 2-bromo-5-chloro, pubchem1169, acmc-209jeb, 2-bromo-5-chloro pyridine, ksc238i0d, tpc-py102, 2-bromanyl-5-chloranyl-pyridine
C1=CC(=NC=C1Cl)Br
192.44
192.44
98%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger