Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-chlorobenzotrifluoride, 98+%

$67.42 - $233.97

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-chlorobenzotrifluoride
CAS 344-65-0
Molecular Formula C7H3BrClF3
Formula Weight 259.44
MDL Number MFCD00010308
Beilstein 2448030
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2013506

 
alfa aesar™
L2013506
5g Each for $67.42

AAL2013514

 
alfa aesar™
L2013514
25g Each for $233.97
Description & Specifications

Specifications

Formula Weight 259.44
Molecular Formula C7H3BrClF3
MDL Number MFCD00010308
CAS 344-65-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 18° to 20°C
Chemical Name or Material 2-Bromo-5-chlorobenzotrifluoride
Assay ≥98%
Quantity 5g
Beilstein 2448030
Boiling Point 198°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.