missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-chlorobenzoic Acid, TCI America™

$74.86 - $264.31

Chemical Identifiers

CAS 21739-93-5
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00013982
InChI Key RBCPJQQJBAQSOU-UHFFFAOYSA-N
Synonym 6-bromo-3-chlorobenzoic acid, benzoic acid, 2-bromo-5-chloro, 2-bromo-5-chloro-benzoic acid, 5-chloro-2-bromobenzoic acid, pubchem3584, acmc-1co7w, 2-bromo-5-chlorobenzoicacid, ksc204k2j, benzoicacid, 2-bromo-5-chloro, zerenex e/9071908
PubChem CID 89027
IUPAC Name 2-bromo-5-chlorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)C(=O)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B31125G View Documents TCI America
B31125G
5g Each for $74.86
 
B311225G View Documents TCI America
B311225G
25g Each for $264.31
 
Specifications

Chemical Identifiers

21739-93-5
235.461
RBCPJQQJBAQSOU-UHFFFAOYSA-N
89027
C1=CC(=C(C=C1Cl)C(=O)O)Br
C7H4BrClO2
MFCD00013982
6-bromo-3-chlorobenzoic acid, benzoic acid, 2-bromo-5-chloro, 2-bromo-5-chloro-benzoic acid, 5-chloro-2-bromobenzoic acid, pubchem3584, acmc-1co7w, 2-bromo-5-chlorobenzoicacid, ksc204k2j, benzoicacid, 2-bromo-5-chloro, zerenex e/9071908
2-bromo-5-chlorobenzoic acid

Specifications

White-Yellow
5g
C7H4BrClO2
2811
RBCPJQQJBAQSOU-UHFFFAOYSA-N
2-bromo-5-chlorobenzoic acid
89027
Crystal-Powder at 20°C
2-Bromo-5-chlorobenzoic Acid
155°C
21739-93-5
MFCD00013982
6-bromo-3-chlorobenzoic acid, benzoic acid, 2-bromo-5-chloro, 2-bromo-5-chloro-benzoic acid, 5-chloro-2-bromobenzoic acid, pubchem3584, acmc-1co7w, 2-bromo-5-chlorobenzoicacid, ksc204k2j, benzoicacid, 2-bromo-5-chloro, zerenex e/9071908
C1=CC(=C(C=C1Cl)C(=O)O)Br
235.461
235.46
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No