missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-chlorobenzoic acid, 98+%

Manufacturer:  Alfa Aesar™ A1010303

 View more versions of this product

Catalog No. AAA1010303



Specifications

Specifications

2-Bromo-5-chlorobenzoic acid
152°C to 156°C
21739-93-5
MFCD00013982
2442261
RBCPJQQJBAQSOU-UHFFFAOYSA-N
2-bromo-5-chlorobenzoic acid
89027
≥98%
Odorless
1g
C7H4BrClO2
UN2811
6-bromo-3-chlorobenzoic acid, benzoic acid, 2-bromo-5-chloro, 2-bromo-5-chloro-benzoic acid, 5-chloro-2-bromobenzoic acid, pubchem3584, acmc-1co7w, 2-bromo-5-chlorobenzoicacid, ksc204k2j, benzoicacid, 2-bromo-5-chloro, zerenex e/9071908
C1=CC(=C(C=C1Cl)C(=O)O)Br
235.461
235.46

Chemical Identifiers

21739-93-5
235.461
RBCPJQQJBAQSOU-UHFFFAOYSA-N
89027
C1=CC(=C(C=C1Cl)C(=O)O)Br
C7H4BrClO2
MFCD00013982
6-bromo-3-chlorobenzoic acid, benzoic acid, 2-bromo-5-chloro, 2-bromo-5-chloro-benzoic acid, 5-chloro-2-bromobenzoic acid, pubchem3584, acmc-1co7w, 2-bromo-5-chlorobenzoicacid, ksc204k2j, benzoicacid, 2-bromo-5-chloro, zerenex e/9071908
2-bromo-5-chlorobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents