missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-chloro-3-nitropyridine, 98%

Manufacturer:  Alfa Aesar™ H6406503

 View more versions of this product

Catalog No. AAH6406503



Specifications

Specifications

2-Bromo-5-chloro-3-nitropyridine
175°C to 179°C
75806-86-9
MFCD07368982
147540
VFMIDVIAQMFGGP-UHFFFAOYSA-N
2-bromo-5-chloro-3-nitropyridine
280681
Solid
White
1g
C5H2BrClN2O2
UN2811
2-bromo-3-nitro-5-chloropyridine, pyridine, 2-bromo-5-chloro-3-nitro, 2-bromo-5-chloro-3-nitro-pyridine, pubchem5951, acmc-1bggp, ksc495s5l, 2-bromo-5-chloro-3-nitro pyridine, 3-nitro-2-bromo-5-chloropyridine, 5-chloro-2-bromo-3-nitropyridine
C1=C(C(=NC=C1Cl)Br)[N+](=O)[O-]
237.437
237.44
98%

Chemical Identifiers

75806-86-9
237.437
VFMIDVIAQMFGGP-UHFFFAOYSA-N
280681
C1=C(C(=NC=C1Cl)Br)[N+](=O)[O-]
C5H2BrClN2O2
MFCD07368982
2-bromo-3-nitro-5-chloropyridine, pyridine, 2-bromo-5-chloro-3-nitro, 2-bromo-5-chloro-3-nitro-pyridine, pubchem5951, acmc-1bggp, ksc495s5l, 2-bromo-5-chloro-3-nitro pyridine, 3-nitro-2-bromo-5-chloropyridine, 5-chloro-2-bromo-3-nitropyridine
2-bromo-5-chloro-3-nitropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents