Please login to your online account to display your discounted pricing

2-Bromo-5-chloro-3-methylbenzo[b]thiophene, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM08172EA

 View more versions of this product

Catalog No. KM08172EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 261.57
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C9H6BrClS
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 2-Bromo-5-chloro-3-methylbenzo[b]thiophene
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00126363
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 175203-60-8

Specifications

Chemical Name or Material 2-Bromo-5-chloro-3-methylbenzo[b]thiophene
CAS 175203-60-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6BrClS
Formula Weight 261.57
MDL Number MFCD00126363
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents