Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-5-chloro-3-methylbenzo(b)thiophene, 97%

$71.58 - $267.84

Chemical Identifiers

Chemical Name or Material 2-Bromo-5-chloro-3-methylbenzo(b)thiophene
CAS 175203-60-8
Molecular Formula C9H6BrClS
Formula Weight 261.57
MDL Number MFCD00126363
Synonym 2-Bromo-5-chloro-3-methylthianaphthene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2025003

  SDS
alfa aesar™
B2025003
1g Each for $71.58

AAB2025006

  SDS
alfa aesar™
B2025006
5g Each for $267.84
Description & Specifications

Specifications

Formula Weight 261.57
Chemical Name or Material 2-Bromo-5-chloro-3-methylbenzo(b)thiophene
Assay 97%
Molecular Formula C9H6BrClS
MDL Number MFCD00126363
CAS 175203-60-8
Melting Point 104° to 107°C
Synonym 2-Bromo-5-chloro-3-methylthianaphthene
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.