Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-(trifluoromethyl)pyridine, 97%

$109.85 - $109.85

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-(trifluoromethyl)pyridine
CAS 175205-81-9
Molecular Formula C6H3BrF3N
Formula Weight 226
MDL Number MFCD00153085
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25748MD

  SDS
alfa aesar™
H25748MD
250mg Each for $109.85
Description & Specifications

Specifications

Formula Weight 226
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)pyridine
Assay 97%
Molecular Formula C6H3BrF3N
MDL Number MFCD00153085
CAS 175205-81-9
Boiling Point 84° to 85°C (14mmHg)
Quantity 250mg
Flash Point 73°C (164°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.