Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-(trifluoromethyl)benzenesulfonyl chloride, 97%


Quantity 5g

Manufacturer: alfa aesar™  L1971206

 View more versions of this product

Catalog No. AAL1971206

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 323.52
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)benzenesulfonyl chloride
Assay 97%
Molecular Formula C7H3BrClF3O2S
CAS 54403-98-4
Boiling Point 235° to 236°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
MDL Number MFCD03094394
UN Number UN3265
Quantity 5g

Specifications

Chemical Name or Material 2-Bromo-4-(trifluoromethyl)benzenesulfonyl chloride
CAS 54403-98-4
Molecular Formula C7H3BrClF3O2S
Formula Weight 323.52
MDL Number MFCD03094394
UN Number UN3265
Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.