Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-(trifluoromethyl)anisole, 97%

$153.16 - $457.16

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-(trifluoromethyl)anisole
CAS 402-10-8
Molecular Formula C8H6BrF3O
Formula Weight 255.04
MDL Number MFCD03789249
Synonym 3-Bromo-4-methoxybenzotrifluoride; 2-Bromo-1-methoxy-4-(trifluoromethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2683806

  SDS
alfa aesar™
H2683806
5g Each for $153.16

AAH2683814

  SDS
alfa aesar™
H2683814
25g Each for $457.16
Description & Specifications

Specifications

Formula Weight 255.04
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)anisole
Assay 97%
Molecular Formula C8H6BrF3O
MDL Number MFCD03789249
CAS 402-10-8
Synonym 3-Bromo-4-methoxybenzotrifluoride; 2-Bromo-1-methoxy-4-(trifluoromethyl)benzene
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.