Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-(trifluoromethyl)aniline
CAS 57946-63-1
Molecular Formula C7H5BrF3N
Formula Weight 240.03
MDL Number MFCD00042150
UN Number UN2811
Beilstein 8139978
Synonym 4-Amino-3-bromobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1852806

  SDS
alfa aesar™
A1852806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1852803

  SDS
alfa aesar™
A1852803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.03
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)aniline
Assay 97%
Molecular Formula C7H5BrF3N
MDL Number MFCD00042150
CAS 57946-63-1
UN Number UN2811
Melting Point 26° to 33°C
Beilstein 8139978
Synonym 4-Amino-3-bromobenzotrifluoride
Boiling Point 109° to 110°C (10mmHg)
Quantity 5g
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.