missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4′-(trifluoromethyl)acetophenone, TCI America™

Manufacturer:  TCI America T274725G

 View more versions of this product

Catalog No. T274725G



Specifications

Specifications

2-Bromo-4′-(trifluoromethyl)acetophenone
55°C
383-53-9
MFCD00126489
2-bromo-1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethyl acetophenone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-4'-trifluoromethyl acetophenon, 4-trifluoromethylphenacyl bromide, ethanone, 2-bromo-1-4-trifluoromethyl phenyl, 4'-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethylacetophenone
C1=CC(=CC=C1C(=O)CBr)C(F)(F)F
267.045
267.05
≥95.0% (GC)
Yellow
25g
C9H6BrF3O
3261
HEMROKPXTCOASZ-UHFFFAOYSA-N
2-bromo-1-[4-(trifluoromethyl)phenyl]ethanone
321979
Crystal-Powder at 20°C

Chemical Identifiers

383-53-9
267.045
HEMROKPXTCOASZ-UHFFFAOYSA-N
321979
C1=CC(=CC=C1C(=O)CBr)C(F)(F)F
C9H6BrF3O
MFCD00126489
2-bromo-1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethyl acetophenone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-4'-trifluoromethyl acetophenon, 4-trifluoromethylphenacyl bromide, ethanone, 2-bromo-1-4-trifluoromethyl phenyl, 4'-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethylacetophenone
2-bromo-1-[4-(trifluoromethyl)phenyl]ethanone
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents