missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-4'-(trifluoromethyl)acetophenone 95.0+%, TCI America™
$117.79 - $486.08
Chemical Identifiers
CAS | 383-53-9 |
---|---|
Molecular Formula | C9H6BrF3O |
Molecular Weight (g/mol) | 267.05 |
MDL Number | MFCD00126489 |
InChI Key | HEMROKPXTCOASZ-UHFFFAOYSA-N |
Synonym | 2-bromo-1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethyl acetophenone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-4'-trifluoromethyl acetophenon, 4-trifluoromethylphenacyl bromide, ethanone, 2-bromo-1-4-trifluoromethyl phenyl, 4'-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethylacetophenone |
PubChem CID | 321979 |
IUPAC Name | 2-bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one |
SMILES | FC(F)(F)C1=CC=C(C=C1)C(=O)CBr |
Chemical Identifiers
383-53-9 | |
267.05 | |
HEMROKPXTCOASZ-UHFFFAOYSA-N | |
321979 | |
FC(F)(F)C1=CC=C(C=C1)C(=O)CBr |
C9H6BrF3O | |
MFCD00126489 | |
2-bromo-1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethyl acetophenone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-4'-trifluoromethyl acetophenon, 4-trifluoromethylphenacyl bromide, ethanone, 2-bromo-1-4-trifluoromethyl phenyl, 4'-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethylacetophenone | |
2-bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one |
Specifications
383-53-9 | |
Yellow | |
MFCD00126489 | |
3261 | |
HEMROKPXTCOASZ-UHFFFAOYSA-N | |
2-bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one | |
321979 | |
≥95.0% (GC) | |
2-Bromo-4′-(trifluoromethyl)acetophenone |
55°C | |
C9H6BrF3O | |
5 g | |
2-bromo-1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethyl acetophenone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-4'-trifluoromethyl acetophenon, 4-trifluoromethylphenacyl bromide, ethanone, 2-bromo-1-4-trifluoromethyl phenyl, 4'-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethylacetophenone | |
FC(F)(F)C1=CC=C(C=C1)C(=O)CBr | |
267.05 | |
267.05 | |
Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator