Please login to your online account to display your discounted pricing

2-Bromo-4-trifluoromethoxyaniline, 98%, ACROS Organics™

$95.68 - $95.68

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-trifluoromethoxyaniline
CAS 175278-17-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H5BrF3NO
Formula Weight 256.02
MDL Number MFCD00203478
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC350170250

  SDS
thermo fisher scientific
AC350170250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350170010

  SDS
thermo fisher scientific
35017-0010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350170050

  SDS
thermo fisher scientific
35017-0050
Glass bottle 5g Each for $95.68
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 256.02
Assay ≥97.5%
Quantity 25g
Color Clear light yellow to orange-yellow
Density 1.693g/mL
CAS Max % 100.0
Chemical Name or Material 2-Bromo-4-trifluoromethoxyaniline
Packaging Glass bottle
Assay Percent Range 98%
Storage Information Product may darken on storage
Molecular Formula C7H5BrF3NO
CAS 175278-17-8
Infrared Spectrum Authentic
CAS Min % 97.5
Refractive Index 1.5030 to 1.5050 (20°C, 589nm)
MDL Number MFCD00203478
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.