missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-pyridinemethanol, 96%

$98.69 - $368.05

Chemical Identifiers

CAS 118289-16-0
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD04039313
InChI Key IQNUGAFIKDRYRP-UHFFFAOYSA-N
Synonym 2-bromo-4-hydroxymethyl pyridine
PubChem CID 14512957
IUPAC Name (2-bromopyridin-4-yl)methanol
SMILES C1=CN=C(C=C1CO)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1985603 View Documents Alfa Aesar™
L1985603
1g Each for $98.69
 
AAL1985606 View Documents Alfa Aesar™
L1985606
5g Each for $368.05
 
Specifications

Chemical Identifiers

118289-16-0
188.024
IQNUGAFIKDRYRP-UHFFFAOYSA-N
14512957
C1=CN=C(C=C1CO)Br
C6H6BrNO
MFCD04039313
2-bromo-4-hydroxymethyl pyridine
(2-bromopyridin-4-yl)methanol

Specifications

59°C to 63°C
118289-16-0
MFCD04039313
2-bromo-4-hydroxymethyl pyridine
C1=CN=C(C=C1CO)Br
188.024
188.02
2-Bromo-4-pyridinemethanol
1g
C6H6BrNO
4244047
IQNUGAFIKDRYRP-UHFFFAOYSA-N
(2-bromopyridin-4-yl)methanol
14512957
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures