Please login to your online account to display your discounted pricing

2-Bromo-4'-phenylacetophenone 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  146350250

 View more versions of this product

Catalog No. AC146350250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 275.13
Quantity 25g
Color Yellow to beige-brown
Beilstein 07, III, 2137
EINECS Number 205-217-6
TSCA TSCA
Melting Point 123° to 127°C
CAS Max % 100.0
Assay Percent Range 98%
Synonym 4-Phenylphenacyl bromide
Infrared Spectrum Authentic
Chemical Name or Material 2-Bromo-4'-phenylacetophenone
Name Note 98%
Molecular Formula C14H11BrO
MDL Number MFCD00000202
CAS 135-73-9
CAS Min % 97.5
Linear Formula C6H5C6H4COCH2Br
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material 2-Bromo-4'-phenylacetophenone
Name Note 98%
CAS 135-73-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H11BrO
Linear Formula C6H5C6H4COCH2Br
Formula Weight 275.13
MDL Number MFCD00000202
Synonym 4-Phenylphenacyl bromide
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.