Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-nitropyridine N-oxide, 97%

$76.39 - $286.28

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-nitropyridine N-oxide
CAS 52092-43-0
Molecular Formula C5H3BrN2O3
Formula Weight 219
MDL Number MFCD00160743
Beilstein 145698
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2002003

  SDS
alfa aesar™
L2002003
1g Each for $76.39

AAL2002006

  SDS
alfa aesar™
L2002006
5g Each for $286.28
Description & Specifications

Specifications

Formula Weight 219
Chemical Name or Material 2-Bromo-4-nitropyridine N-oxide
CAS 52092-43-0
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C5H3BrN2O3
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 145698
Melting Point 143° to 146°C
Quantity 1g
MDL Number MFCD00160743
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.