Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-nitrophenetole, 99%

$87.28 - $738.85

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-nitrophenetole
CAS 58244-42-1
Molecular Formula C8H8BrNO3
Formula Weight 246.06
MDL Number MFCD12547883
Synonym 2-Bromo-1-ethoxy-4-nitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2543306

  SDS
alfa aesar™
B2543306
5g Each for $87.28

AAB2543314

  SDS
alfa aesar™
B2543314
25g Each for $257.05

AAB2543322

  SDS
alfa aesar™
B2543322
100g Each for $738.85
Description & Specifications

Specifications

Formula Weight 246.06
Chemical Name or Material 2-Bromo-4-nitrophenetole
Assay 99%
Molecular Formula C8H8BrNO3
MDL Number MFCD12547883
CAS 58244-42-1
Melting Point 94° to 98°C
Synonym 2-Bromo-1-ethoxy-4-nitrobenzene
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.