Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-nitroanisole, 98%

$57.57 - $174.78

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-nitroanisole
CAS 5197-28-4
Molecular Formula C7H6BrNO3
Formula Weight 232.03
MDL Number MFCD00041242
Beilstein 2556372
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0718106

  SDS
alfa aesar™
L0718106
5g Each for $57.57

AAL0718114

  SDS
alfa aesar™
L0718114
25g Each for $174.78
Description & Specifications

Specifications

Formula Weight 232.03
MDL Number MFCD00041242
Chemical Name or Material 2-Bromo-4-nitroanisole
Assay 98%
Molecular Formula C7H6BrNO3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 103° to 108°C
Beilstein 2556372
Quantity 5g
CAS 5197-28-4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.