missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-methylpyrimidine, 97%

Manufacturer:  Alfa Aesar™ H5450306

 View more versions of this product

Catalog No. AAH5450306



Description

Description

Applications
2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Bromo-4-methylpyrimidine
5g
C5H5BrN2
2-bromo-4-methlypyrimidine, 2-bromo-4-methyl pyrimidine, 2-bromo-4-methyl-pyrimidine, 2-bromo-4-methylpyrimidine, acmc-209z1w, pubchem7007, pyrimidine, 2-bromo-4-methyl, pyrimidine,2-bromo-4-methyl
UZIJEBOLOXOVFY-UHFFFAOYSA-N
2-bromo-4-methylpyrimidine
10942928
97%
45°C to 50°C
130645-48-6
MFCD04037883
Slightly soluble in water.
CC1=NC(=NC=C1)Br
173.013
173.01

Chemical Identifiers

130645-48-6
173.013
UZIJEBOLOXOVFY-UHFFFAOYSA-N
10942928
CC1=NC(=NC=C1)Br
C5H5BrN2
MFCD04037883
2-bromo-4-methlypyrimidine, 2-bromo-4-methyl pyrimidine, 2-bromo-4-methyl-pyrimidine, 2-bromo-4-methylpyrimidine, acmc-209z1w, pubchem7007, pyrimidine, 2-bromo-4-methyl, pyrimidine,2-bromo-4-methyl
2-bromo-4-methylpyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents